The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206822.1 | NP_001193751.1 | 15 - 16 | 2401 | 101 |
GenBank mRNA | BE217572.1 | - | 1 - 2 | 123 | 101 |