Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
540919
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
C11orf30, C15H11orf30
Chromosome Location
Chr.15: 55852825 - 55934631 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206822.1 NP_001193751.1 9 - 10 1403 84

View Full Product Details