Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NTN5
Entrez Gene ID
785970
Gene Name
netrin 5
Gene Aliases
-
Chromosome Location
Chr.18: 55294153 - 55300421 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206872.1 NP_001193801.1 1 - 2 631 107
GenBank mRNA DR712887.1 - 1 - 2 529 107

View Full Product Details