Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSTM5
Entrez Gene ID
100141246
Gene Name
V-set and transmembrane domain containing 5
Gene Aliases
C11orf90, C29H11orf90
Chromosome Location
Chr.29: 975008 - 1003566 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206964.1 NP_001193893.1 3 - 4 559 95

View Full Product Details