Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIGNL2
Entrez Gene ID
518615
Gene Name
fidgetin like 2
Gene Aliases
-
Chromosome Location
Chr.5: 28014763 - 28045338 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001214905.1 NP_001201834.1 1 - 1 819 65

View Full Product Details