Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNALI1
Entrez Gene ID
529053
Gene Name
dynein axonemal light intermediate chain 1
Gene Aliases
-
Chromosome Location
Chr.3: 108330553 - 108338022 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191299.2 NP_001178228.2 1 - 2 125 71

View Full Product Details