Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFI1
Entrez Gene ID
100139952
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
-
Chromosome Location
Chr.3: 50945342 - 50956131 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191312.2 NP_001178241.1 2 - 3 364 49

View Full Product Details