Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLAMF7
Entrez Gene ID
790164
Gene Name
SLAM family member 7
Gene Aliases
-
Chromosome Location
Chr.3: 8848948 - 8865031 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191358.1 NP_001178287.1 6 - 7 974 81
GenBank mRNA HQ728434.1 - 7 - 8 858 81
HQ728435.1 - 5 - 6 833 81
HQ728436.1 - 6 - 7 741 81

View Full Product Details