Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB3D
Entrez Gene ID
100139105
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 15738325 - 15747393 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191375.1 NP_001178304.1 3 - 4 563 67

View Full Product Details