Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NSUN4
Entrez Gene ID
618779
Gene Name
NOP2/Sun RNA methyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.3: 99748813 - 99772626 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191381.1 NP_001178310.1 5 - 6 906 69
GenBank mRNA BT026151.1 - 5 - 6 855 69

View Full Product Details