Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX1
Entrez Gene ID
528169
Gene Name
pannexin 1
Gene Aliases
-
Chromosome Location
Chr.29: 611569 - 670980 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245925.1 NP_001232854.1 2 - 3 315 67
GenBank mRNA HM036582.1 - 2 - 3 315 67

View Full Product Details