Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PABPC1
Entrez Gene ID
282296
Gene Name
poly(A) binding protein cytoplasmic 1
Gene Aliases
PABP-1
Chromosome Location
Chr.14: 63625778 - 63643551 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174568.3 NP_776993.1 13 - 14 2337 69
GenBank mRNA AJ401269.2 - 13 - 14 1823 69
BC102365.1 - 13 - 14 2327 69
DV850814.1 - 2 - 3 157 69

Gene Symbol
PABPC1P10
Entrez Gene ID
282297
Gene Name
poly(A) binding protein, cytoplasmic 1 pseudogene 10
Gene Aliases
PABPL1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA U83076.1 - 2 - 3 296 69

View Full Product Details