Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDA20
Entrez Gene ID
286791
Gene Name
major allergen BDA20
Gene Aliases
-
Chromosome Location
Chr.X: 137182425 - 137188485 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174761.2 NP_777186.1 3 - 4 312 98
GenBank mRNA L42867.1 - 3 - 4 312 98

View Full Product Details