Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHOX
Entrez Gene ID
615159
Gene Name
short stature homeobox
Gene Aliases
-
Chromosome Location
Chr.2: 121446978 - 121458374 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191546.2 NP_001178475.2 1 - 2 285 82

View Full Product Details