Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARRDC4
Entrez Gene ID
538517
Gene Name
arrestin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.21: 8736193 - 8748958 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192034.2 NP_001178963.1 5 - 6 866 90
GenBank mRNA KP731623.1 - 5 - 6 992 90

View Full Product Details