Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRHPR
Entrez Gene ID
504764
Gene Name
glyoxylate and hydroxypyruvate reductase
Gene Aliases
-
Chromosome Location
Chr.8: 61555137 - 61564033 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192037.3 NP_001178966.1 2 - 3 237 77
GenBank mRNA BC120229.1 - 2 - 3 242 77

View Full Product Details