Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2H
Entrez Gene ID
505599
Gene Name
RNA polymerase II subunit H
Gene Aliases
-
Chromosome Location
Chr.1: 82825044 - 82829473 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192065.2 NP_001178994.1 4 - 5 1524 88
GenBank mRNA BC118247.1 - 3 - 4 900 88

View Full Product Details