Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17F
Entrez Gene ID
506030
Gene Name
interleukin 17F
Gene Aliases
-
Chromosome Location
Chr.23: 24645643 - 24653192 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192082.1 NP_001179011.1 2 - 3 305 79

View Full Product Details