Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCK
Entrez Gene ID
506290
Gene Name
IQ motif containing K
Gene Aliases
-
Chromosome Location
Chr.25: 17314228 - 17475638 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192088.3 NP_001179017.1 2 - 3 263 96

View Full Product Details