Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCM1
Entrez Gene ID
317779
Gene Name
glial cells missing transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.23: 25311576 - 25332914 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192105.1 NP_001179034.1 1 - 2 62 96

View Full Product Details