Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSCAN10
Entrez Gene ID
538853
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
-
Chromosome Location
Chr.25: 2540747 - 2544804 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192113.1 NP_001179042.1 4 - 5 619 100

View Full Product Details