Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDKRB2
Entrez Gene ID
538896
Gene Name
bradykinin receptor B2
Gene Aliases
-
Chromosome Location
Chr.21: 61081624 - 61122153 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192126.1 NP_001179055.1 2 - 3 193 58

View Full Product Details