Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS5
Entrez Gene ID
507179
Gene Name
COP9 signalosome subunit 5
Gene Aliases
-
Chromosome Location
Chr.14: 31193333 - 31213028 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192139.1 NP_001179068.1 4 - 5 728 95

View Full Product Details