Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAMALIN
Entrez Gene ID
507181
Gene Name
trafficking regulator and scaffold protein tamalin
Gene Aliases
GRASP
Chromosome Location
Chr.5: 27865985 - 27873788 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192141.1 NP_001179070.1 3 - 4 423 75

View Full Product Details