Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AVPR1B
Entrez Gene ID
282861
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
Chromosome Location
Chr.16: 3876671 - 3882191 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192142.1 NP_001179071.1 1 - 2 941 71

View Full Product Details