Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB46
Entrez Gene ID
507985
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
-
Chromosome Location
Chr.13: 53978514 - 54030117 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192164.1 NP_001179093.1 3 - 4 1395 91

View Full Product Details