Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BOD1L1
Entrez Gene ID
508527
Gene Name
biorientation of chromosomes in cell division 1 like 1
Gene Aliases
BOD1L
Chromosome Location
Chr.6: 108777761 - 108831167 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192190.3 NP_001179119.2 11 - 12 7957 86
GenBank mRNA BC149869.1 - 2 - 3 230 86

View Full Product Details