Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAK6
Entrez Gene ID
539224
Gene Name
p21 (RAC1) activated kinase 6
Gene Aliases
-
Chromosome Location
Chr.10: 35856111 - 35895687 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192198.1 NP_001179127.1 1 - 2 86 102

View Full Product Details