Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNJ1
Entrez Gene ID
539250
Gene Name
potassium inwardly rectifying channel subfamily J member 1
Gene Aliases
-
Chromosome Location
Chr.29: 32212882 - 32244725 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192207.1 NP_001179136.1 2 - 3 368 93

View Full Product Details