Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPAR2
Entrez Gene ID
509748
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
EDG4
Chromosome Location
Chr.7: 3704289 - 3709843 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192235.1 NP_001179164.1 1 - 2 67 55

View Full Product Details