Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALPK2
Entrez Gene ID
510218
Gene Name
alpha kinase 2
Gene Aliases
-
Chromosome Location
Chr.24: 57611963 - 57745147 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192248.1 NP_001179177.1 7 - 8 5526 73

View Full Product Details