Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCL2
Entrez Gene ID
510408
Gene Name
phospholipase C like 2
Gene Aliases
-
Chromosome Location
Chr.1: 153882096 - 154095595 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192255.2 NP_001179184.2 5 - 6 3181 68

View Full Product Details