Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSD1L
Entrez Gene ID
510535
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
-
Chromosome Location
Chr.8: 95270578 - 95341344 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192260.1 NP_001179189.1 2 - 3 243 72

View Full Product Details