Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCN3
Entrez Gene ID
539578
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
Chromosome Location
Chr.3: 15354861 - 15363825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192278.2 NP_001179207.2 3 - 4 878 69

View Full Product Details