Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LZTS1
Entrez Gene ID
539634
Gene Name
leucine zipper tumor suppressor 1
Gene Aliases
-
Chromosome Location
Chr.8: 67361738 - 67428879 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192287.3 NP_001179216.2 2 - 3 1274 96

View Full Product Details