Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX1
Entrez Gene ID
539689
Gene Name
ALX homeobox 1
Gene Aliases
-
Chromosome Location
Chr.5: 14917352 - 14939197 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192291.2 NP_001179220.1 3 - 4 880 71

View Full Product Details