Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BARHL2
Entrez Gene ID
539708
Gene Name
BarH like homeobox 2
Gene Aliases
-
Chromosome Location
Chr.3: 52532079 - 52536695 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192297.1 NP_001179226.1 1 - 2 602 56

View Full Product Details