Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PES1
Entrez Gene ID
511676
Gene Name
pescadillo ribosomal biogenesis factor 1
Gene Aliases
-
Chromosome Location
Chr.17: 69555245 - 69568689 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001354818.1 NP_001341747.1 11 - 12 1250 100
GenBank mRNA BC118072.1 - 10 - 11 2591 100
DV851730.1 - 3 - 4 345 100

View Full Product Details