Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC23
Entrez Gene ID
512177
Gene Name
zinc finger DHHC-type palmitoyltransferase 23
Gene Aliases
-
Chromosome Location
Chr.1: 58619097 - 58633956 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192314.1 NP_001179243.1 1 - 2 136 88

View Full Product Details