Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35D3
Entrez Gene ID
539925
Gene Name
solute carrier family 35 member D3
Gene Aliases
-
Chromosome Location
Chr.9: 74889556 - 74891824 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192332.1 NP_001179261.1 1 - 2 444 80

View Full Product Details