Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FANCE
Entrez Gene ID
513464
Gene Name
FA complementation group E
Gene Aliases
-
Chromosome Location
Chr.23: 9415909 - 9425061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192349.2 NP_001179278.2 5 - 6 1304 57

View Full Product Details