Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKR1D1
Entrez Gene ID
513855
Gene Name
aldo-keto reductase family 1 member D1
Gene Aliases
-
Chromosome Location
Chr.4: 101778260 - 101828908 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192358.1 NP_001179287.1 3 - 4 456 103

View Full Product Details