Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FILIP1
Entrez Gene ID
514193
Gene Name
filamin A interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 14915045 - 15137938 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192362.1 NP_001179291.1 3 - 4 627 93

View Full Product Details