Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAB2
Entrez Gene ID
540203
Gene Name
TGF-beta activated kinase 1 (MAP3K7) binding protein 2
Gene Aliases
MAP3K7IP2
Chromosome Location
Chr.9: 86568903 - 86653592 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192372.1 NP_001179301.1 4 - 5 1947 122

View Full Product Details