Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRT2
Entrez Gene ID
514435
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.8: 43506901 - 43514111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192373.1 NP_001179302.1 2 - 3 964 101
GenBank mRNA BC142311.1 - 2 - 3 965 101

View Full Product Details