Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRP8
Entrez Gene ID
540208
Gene Name
ribosomal RNA processing 8
Gene Aliases
-
Chromosome Location
Chr.15: 46462641 - 46466203 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192374.2 NP_001179303.2 3 - 4 1011 61

View Full Product Details