Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX59
Entrez Gene ID
514901
Gene Name
DEAD-box helicase 59
Gene Aliases
-
Chromosome Location
Chr.16: 79300368 - 79317807 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192389.1 NP_001179318.1 1 - 2 808 103

View Full Product Details