Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPOX
Entrez Gene ID
540283
Gene Name
coproporphyrinogen oxidase
Gene Aliases
-
Chromosome Location
Chr.1: 42888156 - 42901415 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192393.3 NP_001179322.1 5 - 6 1275 74

View Full Product Details