Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH1
Entrez Gene ID
536811
Gene Name
junctophilin 1
Gene Aliases
-
Chromosome Location
Chr.14: 37502881 - 37595438 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192887.3 NP_001179816.1 4 - 5 1897 71

View Full Product Details