Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT10
Entrez Gene ID
537613
Gene Name
synaptotagmin 10
Gene Aliases
-
Chromosome Location
Chr.5: 76301893 - 76417740 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192912.1 NP_001179841.1 4 - 5 1194 70

View Full Product Details